Product Details

SNP ID
rs75081362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:116939748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATCCAACAATGTCAAAGAAACAC[C/T]TGGTTTTCACCCTGAGTCTAAGCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AARD PubMed Links

Gene Details

Gene
AARD
Gene Name
alanine and arginine rich domain containing protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025357.2 Intron NP_001020528.1
XM_011517043.2 Intron XP_011515345.1

View Full Product Details