Product Details

SNP ID
rs78019340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:121639724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCCCGGTTCTCACCCTCATACA[A/G]ACCCTCCTTTCAGACCCGCCCTCCT
Phenotype
MIM: 601636 MIM: 614353
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HAS2 PubMed Links

Gene Details

Gene
HAS2
Gene Name
hyaluronan synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005328.2 Intron NP_005319.1
Gene
HAS2-AS1
Gene Name
HAS2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details