Product Details

SNP ID
rs78171662
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:89908752 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATGCTAAAACTAATTTCTAGAG[A/C]GCCAATTGAGAAAGAAAAATTGGAA
Phenotype
MIM: 604598
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OSGIN2 PubMed Links

Gene Details

Gene
OSGIN2
Gene Name
oxidative stress induced growth inhibitor family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126111.2 Intron NP_001119583.1
NM_004337.2 Intron NP_004328.1
XM_011517287.2 Intron XP_011515589.1
XM_011517288.2 Intron XP_011515590.1

View Full Product Details