Product Details

SNP ID
rs78968776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:139727725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAAACCATCTACAGGTCCGGAC[C/T]TTGCTTGAATACTTCTATTGACAAA
Phenotype
MIM: 611966
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRAPPC9 PubMed Links

Gene Details

Gene
TRAPPC9
Gene Name
trafficking protein particle complex 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160372.3 6890 UTR 3 NP_001153844.1
NM_001321646.1 6890 UTR 3 NP_001308575.1
NM_031466.7 6890 UTR 3 NP_113654.4
XM_011517326.2 6890 Intron XP_011515628.1
XM_011517328.2 6890 Intron XP_011515630.1
XM_011517330.2 6890 Intron XP_011515632.1
XM_017013893.1 6890 Intron XP_016869382.1
XM_017013894.1 6890 Intron XP_016869383.1

View Full Product Details