Product Details
- SNP ID
-
rs78400087
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:142752422 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTCTCCGCAGGCAGCCAGCACCAG[C/G]GCCAGGAGCGCCAGCCGCGTCCCCC
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LYPD2
PubMed Links
Gene Details
- Gene
- LYPD2
- Gene Name
- LY6/PLAUR domain containing 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_205545.2 |
113 |
Silent Mutation |
GCC,GCG |
A10A |
NP_991108.1 |
View Full Product Details