Product Details

SNP ID
rs75630201
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:63169362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCTTCCTTTTTCTCCTCTTTA[C/T]CGCATCTTTCGTCTTGCAACACAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
YTHDF3 PubMed Links

Gene Details

Gene
YTHDF3
Gene Name
YTH N6-methyladenosine RNA binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277813.1 Intron NP_001264742.1
NM_001277814.1 Intron NP_001264743.1
NM_001277815.1 Intron NP_001264744.1
NM_001277816.1 Intron NP_001264745.1
NM_001277817.1 Intron NP_001264746.1
NM_001277818.1 Intron NP_001264747.1
NM_152758.5 Intron NP_689971.4
XM_011517509.2 Intron XP_011515811.1
XM_011517510.2 Intron XP_011515812.1
Gene
YTHDF3-AS1
Gene Name
YTHDF3 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details