Product Details

SNP ID
rs76203594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:73960493 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACAAAAAGCTTTGAGGTGCCTTT[C/T]TTGATATCAACATGACCTCCAGTTG
Phenotype
MIM: 600788
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TCEB1 PubMed Links

Gene Details

Gene
TCEB1
Gene Name
transcription elongation factor B subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204857.1 Intron NP_001191786.1
NM_001204858.1 Intron NP_001191787.1
NM_001204859.1 Intron NP_001191788.1
NM_001204860.1 Intron NP_001191789.1
NM_001204861.1 Intron NP_001191790.1
NM_001204862.1 Intron NP_001191791.1
NM_001204863.1 Intron NP_001191792.1
NM_001204864.1 Intron NP_001191793.1
NM_005648.3 Intron NP_005639.1
XM_011517580.2 Intron XP_011515882.1
XM_011517581.2 Intron XP_011515883.1

View Full Product Details