Product Details
- SNP ID
-
rs79716681
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:81443432 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCATAGATTCTGGTGCACACCACG[C/T]CCTTCATTTTACATTCCTTAAAAAA
- Phenotype
-
MIM: 170715
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PMP2
PubMed Links
Gene Details
- Gene
- PMP2
- Gene Name
- peripheral myelin protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002677.3 |
463 |
Missense Mutation |
GAC,GGC |
D122G |
NP_002668.1 |
View Full Product Details