Product Details

SNP ID
rs78108930
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:81478898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCATAAACTCTCGTGGAAGTGAC[A/G]CCTTTCATGACGCATTCCTAGACAC
Phenotype
MIM: 600434
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FABP4 PubMed Links

Gene Details

Gene
FABP4
Gene Name
fatty acid binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001442.2 437 Silent Mutation GGC,GGT G122G NP_001433.1

View Full Product Details