Product Details

SNP ID
rs80172172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:27547399 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGACTGACCAGTGAAATCTGAA[C/T]TTGTTCCAAGTAATTAGATTTTCTA
Phenotype
MIM: 614260
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C9orf72 PubMed Links

Gene Details

Gene
C9orf72
Gene Name
chromosome 9 open reading frame 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256054.2 2390 UTR 3 NP_001242983.1
NM_018325.4 2390 UTR 3 NP_060795.1
NM_145005.6 2390 Intron NP_659442.2

View Full Product Details