Product Details

SNP ID
rs77087090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:32555528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGTGTATAGGAAAAACAGATAC[C/T]GGCATGAGACACTGACTAGAAGTAA
Phenotype
MIM: 603322 MIM: 609507
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDUFB6 PubMed Links

Gene Details

Gene
NDUFB6
Gene Name
NADH:ubiquinone oxidoreductase subunit B6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199987.1 Intron NP_001186916.1
NM_002493.4 Intron NP_002484.1
NM_182739.2 Intron NP_877416.1
Gene
TOPORS
Gene Name
TOP1 binding arginine/serine rich protein
There are no transcripts associated with this gene.

Gene
TOPORS-AS1
Gene Name
TOPORS antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details