Product Details

SNP ID
rs79284865
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35906522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCATCACCGTCATCCTGGCCAC[A/G]TATCTCATGTGCCGAATGTGGGCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HRCT1 PubMed Links

Gene Details

Gene
HRCT1
Gene Name
histidine rich carboxyl terminus 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039792.1 331 Missense Mutation ATA,GTA I79V NP_001034881.1
Gene
LINC00961
Gene Name
long intergenic non-protein coding RNA 961
There are no transcripts associated with this gene.

View Full Product Details