Product Details
- SNP ID
-
rs74724209
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:37488338 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGAGGAGCCTGGGATTCCCATCCA[A/G]CTGTGCTGCTGATTGACTGAGTGGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
POLR1E
PubMed Links
Gene Details
- Gene
- POLR1E
- Gene Name
- RNA polymerase I subunit E
View Full Product Details