Product Details

SNP ID
rs76978045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:4681351 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAAAGTGTTTTCCCTGAGGCCAT[A/G]TAACTAGTTAGTGGCCAGGTTAAAA
Phenotype
MIM: 610346
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC37L1 PubMed Links

Gene Details

Gene
CDC37L1
Gene Name
cell division cycle 37 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017913.3 Intron NP_060383.2
Gene
CDC37L1-AS1
Gene Name
CDC37L1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details