Product Details

SNP ID
rs79969282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:90021592 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTTAAAATTTCACTGTGCTGAA[A/G]GAGCTAGTACCAACACAAAAATCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR4290 PubMed Links

Gene Details

Gene
MIR4290
Gene Name
microRNA 4290
There are no transcripts associated with this gene.

Gene
MIR4290HG
Gene Name
MIR4290 host gene
There are no transcripts associated with this gene.

View Full Product Details