Product Details
- SNP ID
-
rs76821927
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:90613426 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGTGCGGGCCAAGGCCTCCGCCTC[G/A]CTGCTCTGCACCTCGCGGCTGGGGC
- Phenotype
-
MIM: 607863
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DIRAS2
PubMed Links
Gene Details
- Gene
- DIRAS2
- Gene Name
- DIRAS family GTPase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017594.4 |
792 |
Silent Mutation |
AGC,AGT |
S134S |
NP_060064.2 |
View Full Product Details