Product Details
- SNP ID
-
rs74987546
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:71863974 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAATGTGGTTTGAACAGCACTTTG[A/T]GTATCATTTTCCCATGATGTATTTT
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ABHD17B
PubMed Links
Gene Details
- Gene
- ABHD17B
- Gene Name
- abhydrolase domain containing 17B
There are no transcripts associated with this gene.
View Full Product Details