Product Details

SNP ID
rs77183607
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104617440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGGGCTTCATATACATGAAGAG[T/G]ATGGTCCCATAGAATATTATGACCA
Phenotype
Polymorphism
T/G, Transversion Substitution
Allele Nomenclature
Literature Links
OR13C9 PubMed Links

Gene Details

Gene
OR13C9
Gene Name
olfactory receptor family 13 subfamily C member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001956.1 765 Silent Mutation ATA,ATC I255I NP_001001956.1

View Full Product Details