Product Details

SNP ID
rs78426461
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104617517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCTTTGCTTCTCCCCTCAGAGG[G/A]GTGAATCTTGAGGATGCTGGAAATG
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
OR13C9 PubMed Links

Gene Details

Gene
OR13C9
Gene Name
olfactory receptor family 13 subfamily C member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001956.1 688 Missense Mutation CCC,TCC P230S NP_001001956.1

View Full Product Details