Product Details

SNP ID
rs76727787
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104617699 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATGATTGATGACATTCTTCCTG[C/A]AGAAAGGCAATTGTACTACAAATGT
Phenotype
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
OR13C9 PubMed Links

Gene Details

Gene
OR13C9
Gene Name
olfactory receptor family 13 subfamily C member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001956.1 506 Missense Mutation TGC,TTC C169F NP_001001956.1

View Full Product Details