Product Details

SNP ID
rs76994656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:108867868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCTATACTGGGAAAGGCTTCTT[A/G]AATTACAATTTTGGCTATATCACTT
Phenotype
MIM: 604303 MIM: 603722
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTL7A PubMed Links

Gene Details

Gene
ACTL7A
Gene Name
actin like 7A
There are no transcripts associated with this gene.

Gene
IKBKAP
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318360.1 5625 UTR 3 NP_001305289.1
NM_003640.4 5625 UTR 3 NP_003631.2
XM_011519136.1 5625 Intron XP_011517438.1
XM_011519137.1 5625 Intron XP_011517439.1
XM_017015238.1 5625 Intron XP_016870727.1

View Full Product Details