Product Details
- SNP ID
-
rs76490277
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:109647244 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTTTCTTATCAATGTATGTCAGC[A/C]TGTGTAGCTCAGTGGTAAAGCACAT
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PALM2
PubMed Links
Gene Details
- Gene
- PALM2
- Gene Name
- paralemmin 2
View Full Product Details