Product Details

SNP ID
rs75883468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:72524552 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTATAACAACCCAATGAGGTAAG[C/T]GAGGAATAGTTGTTAACTCTAATAC
Phenotype
MIM: 606706
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMC1 PubMed Links

Gene Details

Gene
TMC1
Gene Name
transmembrane channel like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138691.2 Intron NP_619636.2
XM_017014256.1 Intron XP_016869745.1

View Full Product Details