Product Details

SNP ID
rs80243781
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:111528436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAATTACATATTATAAATACAAAT[A/G]TATAAACTATAAACTGAATTACGGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF483 PubMed Links

Gene Details

Gene
ZNF483
Gene Name
zinc finger protein 483
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007169.3 Intron NP_001007170.1
NM_133464.3 Intron NP_597721.2
XM_011518300.2 Intron XP_011516602.1
XM_017014337.1 Intron XP_016869826.1
XM_017014338.1 Intron XP_016869827.1
XM_017014339.1 Intron XP_016869828.1

View Full Product Details