Product Details

SNP ID
rs76303905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:114093496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGGTAGCAGGCAGAGAGGAGAC[A/G]CCTAGAAAGAACAGCAGGGTCTATG
Phenotype
MIM: 611278
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF12 PubMed Links

Gene Details

Gene
KIF12
Gene Name
kinesin family member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138424.1 1467 Missense Mutation CGT,TGT R330C NP_612433.1
XM_005251683.4 1467 Missense Mutation CGT,TGT R468C XP_005251740.1
XM_006716947.2 1467 Missense Mutation CGT,TGT R434C XP_006717010.1
XM_011518190.2 1467 Missense Mutation CGT,TGT R526C XP_011516492.1
XM_011518191.2 1467 Missense Mutation CGT,TGT R526C XP_011516493.1
XM_011518192.2 1467 Missense Mutation CGT,TGT R526C XP_011516494.1
XM_011518193.2 1467 Intron XP_011516495.1
XM_017014240.1 1467 Missense Mutation CGT,TGT R203C XP_016869729.1

View Full Product Details