Product Details

SNP ID
rs74686266
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:114893971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGAACCGTTTCCTGGCTATGTC[A/G]GTTTAGGCCCAGAGTGACTGGTACC
Phenotype
MIM: 603875
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNFSF8 PubMed Links

Gene Details

Gene
TNFSF8
Gene Name
tumor necrosis factor superfamily member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244.3 895 Intron NP_001235.1
NM_001252290.1 895 UTR 3 NP_001239219.1

View Full Product Details