Product Details

SNP ID
rs75066080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:120602562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCATTGAGAAAAATTATAACCAT[A/G]ATCTAGGAGCCATTAACAAGTATTT
Phenotype
MIM: 604268
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MEGF9 PubMed Links

Gene Details

Gene
MEGF9
Gene Name
multiple EGF like domains 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080497.2 4566 UTR 3 NP_001073966.2

View Full Product Details