Product Details

SNP ID
rs78378194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:129175777 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAACAAAAGGCTTTTAAATTAAA[C/T]AAATCTATCGAGCTGAAGACACAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IER5L PubMed Links

Gene Details

Gene
IER5L
Gene Name
immediate early response 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203434.2 2485 UTR 3 NP_982258.2

View Full Product Details