Product Details
- SNP ID
-
rs77889648
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:129813141 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAATTTAGAATCCAGCAGAGAGCAC[A/G]TGTGGGCTATTTGGAATGGACAGTG
- Phenotype
-
MIM: 605204
MIM: 608050
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TOR1A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3842225] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TOR1A
- Gene Name
- torsin family 1 member A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000113.2 |
1907 |
UTR 3 |
|
|
NP_000104.1 |
- Gene
- TOR1B
- Gene Name
- torsin family 1 member B
There are no transcripts associated with this gene.
View Full Product Details