Product Details

SNP ID
rs76989928
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:74957872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGGGGGAGACAGACAATAAATA[A/G]GCACATCTATCAGGTGGTGCTGAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf40 PubMed Links

Gene Details

Gene
C9orf40
Gene Name
chromosome 9 open reading frame 40
There are no transcripts associated with this gene.

Gene
C9orf41-AS1
Gene Name
C9orf41 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details