Product Details

SNP ID
rs74561802
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:14821613 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGAACCCCTCCTGCACAGGTAC[G/T]GAAGGACAACAGTGTATACAAAGTT
Phenotype
MIM: 611233
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CDNF PubMed Links

Gene Details

Gene
CDNF
Gene Name
cerebral dopamine neurotrophic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029954.2 Intron NP_001025125.2
XM_011519488.2 Intron XP_011517790.1

View Full Product Details