Product Details

SNP ID
rs79400488
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:27115632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGCCTAAAATCATTTTAAGTC[C/T]GGTTCTCAGTACTTTCTTGAAAAAC
Phenotype
MIM: 607472
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
YME1L1 PubMed Links

Gene Details

Gene
YME1L1
Gene Name
YME1 like 1 ATPase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253866.1 Intron NP_001240795.1
NM_014263.3 Intron NP_055078.1
NM_139312.2 Intron NP_647473.1
XM_011519300.2 Intron XP_011517602.1

View Full Product Details