Product Details

SNP ID
rs74652496
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:7819698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTTGATGTGAAGCATTGTATGT[G/T]TGTTTGTTGAGGAGAGGAAACTACC
Phenotype
MIM: 606576
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TAF3 PubMed Links

Gene Details

Gene
TAF3
Gene Name
TATA-box binding protein associated factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031923.3 Intron NP_114129.1
XM_011519741.1 Intron XP_011518043.1

View Full Product Details