Product Details

SNP ID
rs74326890
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:132185539 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCCAGGGTTTCGGAGTCAGGGCT[G/T]TCTTGTCTGGCGTGGGACGGCCCAA
Phenotype
MIM: 608407 MIM: 611198
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DPYSL4 PubMed Links

Gene Details

Gene
DPYSL4
Gene Name
dihydropyrimidinase like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006426.2 Intron NP_006417.2
XM_005252658.3 Intron XP_005252715.1
XM_011539125.2 Intron XP_011537427.1
XM_017015482.1 Intron XP_016870971.1
XM_017015483.1 Intron XP_016870972.1
Gene
JAKMIP3
Gene Name
Janus kinase and microtubule interacting protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105521.3 Intron NP_001098991.1
NM_001323086.1 Intron NP_001310015.1
NM_001323087.1 Intron NP_001310016.1
NM_001323088.1 Intron NP_001310017.1
NM_001323089.1 Intron NP_001310018.1
NM_001323090.1 Intron NP_001310019.1
XM_005252674.4 Intron XP_005252731.1
XM_005252676.4 Intron XP_005252733.1
XM_011539676.2 Intron XP_011537978.1
XM_011539682.2 Intron XP_011537984.1
XM_017016095.1 Intron XP_016871584.1
XM_017016096.1 Intron XP_016871585.1
XM_017016097.1 Intron XP_016871586.1
XM_017016098.1 Intron XP_016871587.1
XM_017016099.1 Intron XP_016871588.1

View Full Product Details