Product Details

SNP ID
rs77502726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1578694 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGAAGTTGCAGCTCAGCTGGCCA[C/T]GTCTGCACACCCTCTCCCACGAGAC
Phenotype
MIM: 602038 MIM: 148022
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP8 PubMed Links

Gene Details

Gene
DUSP8
Gene Name
dual specificity phosphatase 8
There are no transcripts associated with this gene.

Gene
KRTAP5-1
Gene Name
keratin associated protein 5-1
There are no transcripts associated with this gene.

Gene
KRTAP5-AS1
Gene Name
KRTAP5-1/KRTAP5-2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details