Product Details

SNP ID
rs76831336
Assay Type
Functionally Tested
NCBI dbSNP Submissions
12
Location
Chr.11:26669155 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTATTCTTTCTGTCTCTCTCTTC[C/T]TCTTCCTGTCTTTTTCTTTCTTTCT
Phenotype
MIM: 610110 MIM: 612455
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ANO3 PubMed Links

Gene Details

Gene
ANO3
Gene Name
anoctamin 3
There are no transcripts associated with this gene.

Gene
SLC5A12
Gene Name
solute carrier family 5 member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178498.3 4043 UTR 3 NP_848593.2
XM_006718155.3 4043 UTR 3 XP_006718218.1
XM_006718156.3 4043 Intron XP_006718219.1
XM_011519920.2 4043 UTR 3 XP_011518222.1
XM_011519921.2 4043 Intron XP_011518223.1
XM_017017244.1 4043 UTR 3 XP_016872733.1

View Full Product Details