Product Details

SNP ID
rs79626653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:27338523 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAATATAAAACAATACAAGAAAAA[C/T]TGTTAGTTTTATTGGTATTACAGTG
Phenotype
MIM: 612324
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC34 PubMed Links

Gene Details

Gene
CCDC34
Gene Name
coiled-coil domain containing 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030771.1 1474 UTR 3 NP_110398.1
NM_080654.2 1474 Intron NP_542385.1

View Full Product Details