Product Details
- SNP ID
-
rs78572035
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:27657926 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATCCATGGTAAGGGCCCGCACGTA[T/C]GACTGGGTAGTTCGGCACTGGGAGT
- Phenotype
-
MIM: 113505
MIM: 611468
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BDNF
PubMed Links
Gene Details
- Gene
- BDNF
- Gene Name
- brain derived neurotrophic factor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001143805.1 |
1724 |
Silent Mutation |
TCA,TCG |
S213S |
NP_001137277.1 |
NM_001143806.1 |
1724 |
Silent Mutation |
TCA,TCG |
S213S |
NP_001137278.1 |
NM_001143807.1 |
1724 |
Silent Mutation |
TCA,TCG |
S213S |
NP_001137279.1 |
NM_001143808.1 |
1724 |
Silent Mutation |
TCA,TCG |
S213S |
NP_001137280.1 |
NM_001143809.1 |
1724 |
Silent Mutation |
TCA,TCG |
S242S |
NP_001137281.1 |
NM_001143810.1 |
1724 |
Silent Mutation |
TCA,TCG |
S295S |
NP_001137282.1 |
NM_001143811.1 |
1724 |
Silent Mutation |
TCA,TCG |
S213S |
NP_001137283.1 |
NM_001143812.1 |
1724 |
Silent Mutation |
TCA,TCG |
S213S |
NP_001137284.1 |
NM_001143813.1 |
1724 |
Silent Mutation |
TCA,TCG |
S213S |
NP_001137285.1 |
NM_001143814.1 |
1724 |
Silent Mutation |
TCA,TCG |
S213S |
NP_001137286.1 |
NM_001143816.1 |
1724 |
Silent Mutation |
TCA,TCG |
S213S |
NP_001137288.1 |
NM_001709.4 |
1724 |
Silent Mutation |
TCA,TCG |
S213S |
NP_001700.2 |
NM_170731.4 |
1724 |
Silent Mutation |
TCA,TCG |
S221S |
NP_733927.1 |
NM_170732.4 |
1724 |
Silent Mutation |
TCA,TCG |
S213S |
NP_733928.1 |
NM_170733.3 |
1724 |
Silent Mutation |
TCA,TCG |
S213S |
NP_733929.1 |
NM_170734.3 |
1724 |
Silent Mutation |
TCA,TCG |
S228S |
NP_733930.1 |
NM_170735.5 |
1724 |
Silent Mutation |
TCA,TCG |
S213S |
NP_733931.1 |
XM_011520280.2 |
1724 |
Silent Mutation |
TCA,TCG |
S295S |
XP_011518582.1 |
- Gene
- BDNF-AS
- Gene Name
- BDNF antisense RNA
There are no transcripts associated with this gene.
View Full Product Details