Product Details

SNP ID
rs78845938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:30386560 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTTATAAAATACCCAGTCTCAG[A/G]GATTCCTTTATGGCAGCAAAAACAG
Phenotype
MIM: 600911
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MPPED2 PubMed Links

Gene Details

Gene
MPPED2
Gene Name
metallophosphoesterase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145399.1 3593 UTR 3 NP_001138871.1
NM_001584.2 3593 Intron NP_001575.1
XM_005253110.3 3593 Intron XP_005253167.1
XM_005253111.2 3593 Intron XP_005253168.1
XM_005253112.1 3593 Intron XP_005253169.1
XM_005253114.3 3593 Intron XP_005253171.1
XM_011520346.1 3593 Intron XP_011518648.1
XM_011520347.2 3593 Intron XP_011518649.1
XM_017018231.1 3593 Intron XP_016873720.1
XM_017018232.1 3593 UTR 3 XP_016873721.1
XM_017018233.1 3593 Intron XP_016873722.1
XM_017018234.1 3593 Intron XP_016873723.1
XM_017018235.1 3593 Intron XP_016873724.1
XM_017018236.1 3593 Intron XP_016873725.1
XM_017018237.1 3593 Intron XP_016873726.1

View Full Product Details