Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001166692.1 | 7182 | Intron | NP_001160164.1 | ||
XM_017017053.1 | 7182 | Intron | XP_016872542.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000611.5 | 7182 | UTR 3 | NP_000602.1 | ||
NM_001127223.1 | 7182 | UTR 3 | NP_001120695.1 | ||
NM_001127225.1 | 7182 | UTR 3 | NP_001120697.1 | ||
NM_001127226.1 | 7182 | UTR 3 | NP_001120698.1 | ||
NM_001127227.1 | 7182 | UTR 3 | NP_001120699.1 | ||
NM_203329.2 | 7182 | UTR 3 | NP_976074.1 | ||
NM_203330.2 | 7182 | UTR 3 | NP_976075.1 | ||
NM_203331.2 | 7182 | UTR 3 | NP_976076.1 |