Product Details

SNP ID
rs78153268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:35948132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGAATTGTAGTTACATGGGGGTTC[A/G]TTTTCATTCTTTGAGCTCTAAATTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LDLRAD3 PubMed Links

Gene Details

Gene
LDLRAD3
Gene Name
low density lipoprotein receptor class A domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304263.1 Intron NP_001291192.1
NM_001304264.1 Intron NP_001291193.1
NM_174902.3 Intron NP_777562.1

View Full Product Details