Product Details

SNP ID
rs75607370
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:45241847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTCTAAAACATTTCATAGGAAG[C/G]CACTAACAAGTGCTTTGCCTACACA
Phenotype
MIM: 616347 MIM: 607716
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRDM11 PubMed Links

Gene Details

Gene
PRDM11
Gene Name
PR domain 11
There are no transcripts associated with this gene.

Gene
SYT13
Gene Name
synaptotagmin 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001247987.1 3830 UTR 3 NP_001234916.1
NM_020826.2 3830 UTR 3 NP_065877.1

View Full Product Details