Product Details

SNP ID
rs78206553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:48324961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCATGCAATTAGTTCTATTACT[C/T]ATGTTTCTCCTTGTCTTTATAGGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR4C3 PubMed Links

Gene Details

Gene
OR4C3
Gene Name
olfactory receptor family 4 subfamily C member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004702.1 Intron NP_001004702.1

View Full Product Details