Product Details

SNP ID
rs78490540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:48325129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTTTGCTGATCTATGTGGTCAC[A/G]GTTTGTGGCAACATGCTCATTGTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4C3 PubMed Links

Gene Details

Gene
OR4C3
Gene Name
olfactory receptor family 4 subfamily C member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004702.1 Intron NP_001004702.1

View Full Product Details