Product Details
- SNP ID
-
rs75307540
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:6431659 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTCACCTGCCATGATATGGAGCC[A/G]AGAAGACCCAGGGCAGATAAAGGCC
- Phenotype
-
MIM: 142290
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HPX
PubMed Links
Gene Details
- Gene
- HPX
- Gene Name
- hemopexin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000613.2 |
1172 |
Missense Mutation |
CGG,TGG |
R371W |
NP_000604.1 |
View Full Product Details