Product Details

SNP ID
rs78102086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:26940177 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTACTGACTCCTTAGCCTGTAG[A/G]TTAGAAATAACTGGTTTAGATTAGG
Phenotype
MIM: 615079 MIM: 608858
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASUN PubMed Links

Gene Details

Gene
ASUN
Gene Name
asunder, spermatogenesis regulator
There are no transcripts associated with this gene.

Gene
FGFR1OP2
Gene Name
FGFR1 oncogene partner 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171887.1 Intron NP_001165358.1
NM_001171888.1 Intron NP_001165359.1
NM_015633.2 Intron NP_056448.1

View Full Product Details