Product Details

SNP ID
rs77303541
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:7306698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCGTTTGGCCGTGATTCTGCCC[A/C]GAATCCCTGAGTGGTGGCTGGTCAA
Phenotype
MIM: 614360
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACSM4 PubMed Links

Gene Details

Gene
ACSM4
Gene Name
acyl-CoA synthetase medium-chain family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080454.1 367 Silent Mutation AGA,CGA R123R NP_001073923.1

View Full Product Details