Product Details

SNP ID
rs76754378
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10825579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAAGATAAAGAGGATGATGAAAG[A/G]TATCAAAACTTTCATTGCCTTCACA
Phenotype
MIM: 604791
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TAS2R10 PubMed Links

Gene Details

Gene
TAS2R10
Gene Name
taste 2 receptor member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023921.1 691 Missense Mutation CCT,TCT P231S NP_076410.1

View Full Product Details