Product Details

SNP ID
rs78191133
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10847153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCAGGCTGACAGAAGGAAATCG[C/G]GGTAGAGAGAGTTGACGGTGTCCTC
Phenotype
MIM: 605359
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRH1-PRR4 PubMed Links

Gene Details

Gene
PRH1-PRR4
Gene Name
PRH1-PRR4 readthrough
There are no transcripts associated with this gene.

Gene
PRR4
Gene Name
proline rich 4 (lacrimal)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098538.2 354 Intron NP_001092008.2
NM_007244.2 354 Silent Mutation CCC,CCG P105P NP_009175.2

View Full Product Details