Product Details

SNP ID
rs75436100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:13372925 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCAGCCTGGAAAAATCAAAATGT[A/G]TAGGTAAAGTGAGCACAAGATTTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01559 PubMed Links

Gene Details

Gene
LINC01559
Gene Name
long intergenic non-protein coding RNA 1559
There are no transcripts associated with this gene.

View Full Product Details